missing translation for 'onlineSavingsMsg'
Learn More

(Furfurylthio)acetone, 96%, Thermo Scientific™

Product Code. 11475377
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475377 5 g 5g
11485377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475377 Supplier Thermo Scientific Alfa Aesar Supplier No. B22724.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58066-86-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD02683098
InChI Key WWWHLSMCOBIZNV-UHFFFAOYSA-N
Synonym furfurylthio acetone, 1-furan-2-ylmethylsulfanyl propan-2-one, 1-furfurylthio-2-propanone, 1-furan-2-ylmethyl sulfanyl propan-2-one, 1-2-furylmethylsulfanyl propan-2-one, 2-propanone,1-2-furanylmethyl thio, 2-propanone, 1-2-furanylmethyl thio, 1-2-furylmethyl sulfanyl acetone, furfurythioacetone, furfurylthioacetone
PubChem CID 226249
IUPAC Name 1-(furan-2-ylmethylsulfanyl)propan-2-one
SMILES CC(=O)CSCC1=CC=CO1

Spécification

Quantity 5 g
Formula Weight 170.23
Percent Purity 96%
Chemical Name or Material (Furfurylthio)acetone
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.