Learn More
Carbolite™ FuRNAce TZF12/100/301 tube with 301PID controller 230V 50/60Hz n/a 1200°C/3100W 975mm x 360mm x 525mm Carbolite
Product Code. 12933751
Change view
Click to view available options
Type:
Tube Furnace
Unit Size:
Each
Product Code. Type Unit Size
12933751 Tube Furnace Each
1 options
Product Code. 12933751 Supplier Carbolite™ Supplier No. T12100900230SN+&02301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Tube furnaces provide enhanced thermal uniformity and with the addition of a ceramic work tube and seals, the flexibility to allow materials to be tested under various atmospheric conditions.Typical applications include research tests on inorganic materials, which can be cost effectively carried out in air or in a non-oxidising atmosphere; such as nitrogen or argon or in a vacuum.The enhanced thermal uniformity is ideal for thermocouple calibration and single zone or three zone design are available. Please enquire for more details of the tube furnace range, as there are more sizes and types than this space allows.
  • Symmetrical heating element design ensures improved thermal uniformity compared to chamber furnaces
  • Tube furnace range can accept work tubes from 20mm to 150mm in diameter
  • Seven heated lengths from 300mm to 1,200mm are available
  • Horizontal base mounting is normal, but vertical and split designs are available, with a separate control box
Specifications
Type Tube Furnace