missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Furosemide
Description
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) symporter. Also acts as a non-competitive antagonist at GABAA receptors with ∽ 100-fold greater selectivity for α6-containing receptors than α1-containing receptors.
Specifications
Specifications
| CAS | 54-31-9 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C12H11ClN2O5S |
| Formula Weight | 330.74 |
| Purity | >99% |
| Molecular Weight (g/mol) | 330.739 |
| Chemical Name or Material | Furosemide |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?