missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Fused Silica Capillary Tubing Kit

Product Code. 12432080 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
0.1 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
12432080 0.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12432080 Supplier Cytiva Supplier No. 18114713
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

For Use With For 1/16 in. O.D. Tubing High Pressure Columns (at Low Flow Rates)
Type Capillary Tubing Kit
Includes Kit Contents: 2 x Tubing L= 350 mm, 1 x Tubing L = 800 mm
Diameter (Metric) Inner 0.1 mm
Material Fused Silica
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.