missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ FX-PEGASUS-90 Recirculating Air Safety Storage Cabinets

Product Code. 15664897 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
Drawers 60 kg, Max load 600 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity Unit Size
15664897 Drawers 60 kg, Max load 600 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15664897 Supplier Asecos™ Supplier No. 3291004033627
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the safe, approved and recirculated air filtered storage of flammable liquids and further hazardous substances.

  • Safety for employees – avoidance of explosive atmospheres and extraction and filtering of vapors that are harmful to health
  • Provides flexibility in installation place, storage and interior equipment
  • Enables the common storage, extraction and filtration of the most diverse hazardous materials in one cabinet
  • Electronic monitoring of filter saturation by solvent vapors, air recirculation rate, service intervals and scheduling for the manual filter saturation check
  • Modern bicolor design
  • Eco-friendly with longer filter lifetimes and reusable filter system
  • Fire resistant up to 90 minutes
  • Available interior equipment includes drawers, height-adjustable shelves and bottom collecting sumps
  • Door lockable with cylinder locking, with locking status indicator (red/green)
  • In accordance with EN 14470-1 and TRGS 510
  • Serial automatic door closing releases arrested doors after approx. 60 seconds with a visual and acoustic signal beforehand
  • Recirculating air filter system ideal for the direct installation in working areas with very low noise, only approx. 39 dB
  • With multi-stage wideband filter
  • Monitoring electronics include cool-device cable and power plug
  • Display with innovative touch panel

Specifications

Certifications/Compliance EN 14470-1, TRGS 510
Configuration 4 x 60 kg/19 L powder-coated sheet steel Drawers
Dimensions (D x W x H) Interior 522 x 750 x 1647 mm
Dimensions (D x W x H) 615 x 893 x 2294 mm
Door Style Wing
For Use With (Application) Storage of flammable liquids and other hazardous substances
Load Bearing Capacity Drawers 60 kg, Max load 600 kg
No. of Doors 2
Weight (Metric) 475 kg
Color Anthracite Gray, Warning Yellow
Material Gypsum
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.