missing translation for 'onlineSavingsMsg'
Learn More

G-418 disulfate, MedChemExpress

Product Code. 16479867
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479867 5 g 5g
16421699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479867 Supplier Medchem Express Supplier No. HY17561
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells.

G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells.

Specifications

Chemical Name or Material G-418 disulfate
CAS 108321-42-2
Color White
Purity Grade Notes Research
Molecular Formula C20H44N4O18S2
Quantity 5 g
Synonym Geneticin sulfate Antibiotic G-418 sulfate
Solubility Information H2O : 125 mg/mL (180.45 mM; Need ultrasonic)
SMILES O=S(O)(O)=O.O[C@@H]([C@@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)N)([H])O[C@]2([H])[C@H](O)C)[C@H]([C@@H](C[C@@H]1N)N)O[C@](OC[C@](C)(O)[C@@H]3NC)([H])[C@@H]3O.O=S(O)(O)=O
Molecular Weight (g/mol) 692.71
Formula Weight 692.71
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.