missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO)

Product Code. 11306658
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11306658

Brand: Thermo Scientific Alfa Aesar 013207.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38245-36-2 | C3Gd2O9 | 494.52 g/mol

Gadolinium(III) carbonate is used for making optical glass and dopant for Gadolinium Yttrium Garnets which have microwave applications. It is also used for making phosphors for color TV tube and energy-saving lamps. It is used as intravenous MRI contrast agent to enhance images in medical magnetic resonance imaging and Magnetic Resonance Angiography (MRA) procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-36-2
Molecular Formula C3Gd2O9
Molecular Weight (g/mol) 494.52
MDL Number MFCD00150240
InChI Key RQXZRSYWGRRGCD-UHFFFAOYSA-H
Synonym gadolinium carbonate hydrate, gadolinium iii carbonate hydrate, acmc-1aeff, 3co3.2gd.h2o, gadolinium carbonate-water 2/3/1, digadolinium 3+ hydrate tricarbonate, gadolinium iii carbonate hydrate, reacton?, gadolinium iii carbonate hydrate-gd reo puratrem
PubChem CID 16212943
IUPAC Name digadolinium(3+) tricarbonate
SMILES [Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 494.53 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Assay (REO)
Chemical Name or Material Gadolinium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.