missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) isopropoxide, 99.9% (metals basis), 5% w/v in toluene/isopropanol (3:2)

Product Code. 11329967
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329967 25 mL 25mL
11319967 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329967 Supplier Thermo Scientific Alfa Aesar Supplier No. 042277.AC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Precursor for Gd2O3 films using Sol-Gel process | CAS: 14532-05-9 | C9H21GdO3 | 334.51 g/mol

It is employed in the catalyst action which is used in a ring opening of meso-aziridines with trimethylsilyl azide. In many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14532-05-9
Molecular Formula C9H21GdO3
Molecular Weight (g/mol) 334.51
MDL Number MFCD00144381
InChI Key VJLSFXQJAXVOEQ-UHFFFAOYSA-N
Synonym gadolinium iii isopropoxide, triisopropoxygadolinium, gadolinium isopropoxide, acmc-20akue, gadolinium triisopropoxide, gadolinium tripropan-2-olate, gadolinium-i-propoxide, gadolinium 3+ ion tris propan-2-olate, gadolinium 3+ tris propan-2-olate
PubChem CID 15047187
IUPAC Name gadolinium(3+);propan-2-olate
SMILES [Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Specifications

Color Yellow
Odor Aromatic
Linear Formula Gd[OCH(CH3)2]3
Quantity 25 mL
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes slowly in water.
Formula Weight 334.51
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol (3:2)
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Gadolinium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.