missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxalate hydrate, 99.9% (REO), Thermo Scientific™

Product Code. 11387118
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11387118

Brand: Thermo Scientific Alfa Aesar 015308.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100655-00-3
Molecular Formula C6H8Gd2O13
Molecular Weight (g/mol) 602.62
MDL Number MFCD00150145
InChI Key FEIDMMDEDUAFJQ-UHFFFAOYSA-N
Synonym oxalic acid-gadolinium-water 3/2/1
PubChem CID 92020445
IUPAC Name gadolinium;oxalic acid;hydrate
SMILES O.[Gd].[Gd].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O

Specifications

Boiling Point (decomposition)
Melting Point 110°C (in -6H2O)
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Gd2(C2O4)3·xH2O
UN Number UN3288
Formula Weight 578.56 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Gadolinium(III) oxalate hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.