missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, 99.9%, (trace metal basis)

Product Code. 10067542 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
Product Code. Quantity Packaging unitSize
10067542 10 g Glass bottle 10g
10195900 25 g Glass bottle 25g
10127763 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10067542

Brand: Thermo Scientific Acros 315510100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 1900.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.90%
Linear Formula Gd2O3
Merck Index 10, 4200
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acids
IUPAC Name gadolinium(3+);oxygen(2-)
Formula Weight 362.5
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.