missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium (III) Oxide, TRC

Product Code. 30240936
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240936 10 g 10g
30240644 5 g 5g
30240993 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240936 Supplier Toronto Research Chemicals Supplier No. TRCG12575310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.5
Synonym Digadolinium Trioxide, Gadolinia, Gadolinium Oxide, Gadolinium Oxide (GdO1.5), Gadolinium Sesquioxide, Gadolinium Trioxide, Gadolinium (3+) Oxide
IUPAC Name oxo(oxogadoliniooxy)gadolinium
SMILES O=[Gd]O[Gd]=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/2Gd.3O
Formula Weight 363.8329
Chemical Name or Material Gadolinium (III) Oxide
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.