missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium (III) Oxide, TRC

Product Code. 30240993
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240993 25 g 25g
30240936 10 g 10g
30240644 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240993 Supplier Toronto Research Chemicals Supplier No. TRCG12575325G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.5
Synonym Digadolinium Trioxide, Gadolinia, Gadolinium Oxide, Gadolinium Oxide (GdO1.5), Gadolinium Sesquioxide, Gadolinium Trioxide, Gadolinium (3+) Oxide
IUPAC Name oxo(oxogadoliniooxy)gadolinium
SMILES O=[Gd]O[Gd]=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/2Gd.3O
Formula Weight 363.8329
Chemical Name or Material Gadolinium (III) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.