missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium sputtering target, 76.2mm dia x 3.18mm thick

Product Code. 15497125
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497125 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15497125 Supplier Thermo Scientific Alfa Aesar Supplier No. 045929.KS
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-54-2 | Gd | 157.25 g/mol

Gadolinium is use in neutron radiography and in shielding of nuclear reactors. Used in nuclear marine propulsion systems, used for decarboxylation of oxaloacetic acid, convertion of orto- to para-hydrogen and polymerization of ethylene. It is also used for making gadolinium yttrium, fabrication, color TV tubes, compact discs and computer memory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7440-54-2
Molecular Formula Gd
MDL Number MFCD00011022
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
Solubility Information Soluble in magnesium and diluted acid. Insoluble in water.
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES [Gd]
IUPAC Name gadolinium
Molecular Weight (g/mol) 157.25
PubChem CID 23982
ChEBI CHEBI:33375
Chemical Name or Material Gadolinium sputtering target, 76.2mm dia. x 3.18mm thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.