missing translation for 'onlineSavingsMsg'
Learn More

Galactaric Acid, TRC

Product Code. 30406560
Change view
Click to view available options
Quantity:
50 g
500 g
Unit Size:
500g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406560 50 g 50g
30392414 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406560 Supplier Toronto Research Chemicals Supplier No. TRCG15500550G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mucic acid
CAS 526-99-8
Molecular Formula C6 H10 O8
Quantity 50 g
Synonym Galactaric acid, Galactosaccharic acid, Saccharolactic acid
InChI Formula InChI=1S/C6H10O8/c7-1(3(9)5(11)12)2(8)4(10)6(13)14/h1-4,7-10H,(H,11,12)(H,13,14)
SMILES OC(C(O)C(O)C(=O)O)C(O)C(=O)O
IUPAC Name 2,3,4,5-tetrahydroxyhexanedioic acid
Molecular Weight (g/mol) 210.14
Formula Weight 210.0376
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.