missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacto-Light Plus™ β-Galactosidase Reporter Gene Assay System

Product Code. 12694265
Change view
Click to view available options
Quantity:
15,000 Assays
200 Assays
600 Assays
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12694265 15,000 Assays Each
12684265 200 Assays Each
12614275 600 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12694265 Supplier Invitrogen™ Supplier No. T1009
  • ONLINE EXCLUSIVE PRICE
    6930.00 €
    5613.30 € / 15000 assays
    Save 1316.70 €
    19% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lysis solution, substrate, reaction buffer and accelerator

Rapid, ultrasensitive detection of β-galactosidase in reporter gene assays. Thermo Scientific Galacto-Light Plus β-Galactosidase Reporter Gene Assay System is designed for luminometers without injectors.

The Galacto-Light Plus™ Beta-Galactosidase Reporter Gene Assay System is designed for the rapid, ultrasensitive detection of β-galactosidase in reporter gene assays.

  • The Galacto-Light Plus™ Beta-Galactosidase Reporter Gene Assay System is designed for luminometers without injectors.
  • Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range.
  • Assay sensitivity is 100- to 1,000-fold better than with either the isotopic/non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric/fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays.
  • Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions.
  • Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes.
  • Assay can be completed in about one hour, providing fast assay turnaround.

Sensitive

This chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Ready-to-Use

The Galacto-Light™ Plus™ system incorporates chemiluminescent substrates, reaction buffer, and light-emission accelerator containing luminescence enhancers, in a ready-to-use format. The reaction buffer is supplied at a pH that aids in discrimination of endogenous β-galactosidase, while having little effect on the bacterial form of the enzyme. This enables sensitive detection even in cell lines with relatively high levels of endogenous β-galactosidase activity.

Products described herein are guaranteed for 12 months from the date of purchase.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Chemiluminescence
For Use With (Equipment) Luminometer
Assay β-Gal (lacZ) Assay
Content And Storage This kit contains the lysis solution, substrate, reaction buffer, and accelerator.
Store at 4°C.
Compatible Cells Mammalian Cells
High-throughput Compatibility High-throughput Compatible
Label or Dye Unconjugated
Readout End Point
Sample Type Cell Cultures
Substrate Galacton-Plus™
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Product Line Galacto-Light Plus, NovaBright
For Use With (Application) Enzyme Assay
Quantity 15,000 Assays
Form Liquid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.