missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galacto-Light™ Reaction Buffer Diluent with Galacton-Plus™ Substrate
Click to view available options
Quantity:
1800 assays
Unit Size:
Each
Description
Includes
Concentrated substrate and dilution buffer
For use with Galacto-Light Plus Reporter Gene Assay System.
Specifications
Specifications
| Product Type | ELISA Substrate and Diluent |
| Quantity | 1800 assays |
| Detection Method | Chemiluminescence |
| Conjugate | Unconjugated |
| Form | Liquid |
| Format | 96-well Plates |
| For Use With (Application) | Enzyme Assay |
| For Use With (Equipment) | Luminometer (Microplate) |
| High-throughput Compatibility | High-throughput Compatible |
| Product Line | Galacto-Light, Galacton-Plus |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction