missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)

Product Code. 12654345
Click to view available options
Quantity:
4 mL
10 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12654345

Brand: Invitrogen™ T2266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Concentrated chemiluminescent substrate for E.coli β-galactosidase assays. Thermo Scientific Galacton-Star Substrate (50X Concentrate) is highly sensitive which makes it ideal for detection of weak expression and for transfection normalization with o

Galacton-Star substrate is a chemiluminescent substrate designed for the rapid, and sensitive detection of beta-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric beta-galactosidase assays. Light emission from Galacton-Star reaches maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer. As few as 10 femtograms of beta-galactosidase (20,000 molecules) are detectable.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Substrate
Quantity 10 mL
Detection Method Chemiluminescence
Form Liquid
Format Bottle
Product Line Galacto-Star, Galacton-Star, NovaBright
Substrate Type Beta-Gal
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.