missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)
Click to view available options
Quantity:
4 mL
10 mL
Unit Size:
Each
Description
Galacton-Star substrate is a chemiluminescent substrate designed for the rapid, and sensitive detection of beta-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric beta-galactosidase assays. Light emission from Galacton-Star reaches maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer. As few as 10 femtograms of beta-galactosidase (20,000 molecules) are detectable.
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 10 mL |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Format | Bottle |
| Product Line | Galacto-Star, Galacton-Star, NovaBright |
| Substrate Type | Beta-Gal |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction