missing translation for 'onlineSavingsMsg'
Learn More

Galangin, TRC

Product Code. 30405398
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405398 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405398 Supplier Toronto Research Chemicals Supplier No. TRCG18810025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Galangin
CAS 548-83-4
Purity Grade Notes HPLC
Molecular Formula C15 H10 O5
Quantity 25 mg
Synonym Flavone, 3,5,7-trihydroxy- (7CI,8CI), Galangin (6CI), 3,5,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one, 3,5,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 3,5,7-Trihydroxy-2-phenylchromen-4-one, 3,5,7-Trihydroxyflavone, NSC 407229, Norizalpinin, Galangin
InChI Formula InChI=1S/C15H10O5/c16-9-6-10(17)12-11(7-9)20-15(14(19)13(12)18)8-4-2-1-3-5-8/h1-7,16-17,19H
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccccc3
IUPAC Name 3,5,7-trihydroxy-2-phenylchromen-4-one
Molecular Weight (g/mol) 270.24
Formula Weight 270.0528
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.