missing translation for 'onlineSavingsMsg'
Learn More

Galectin-13 Mouse, Clone: PP13/1161, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 29124-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960128

  • 485.00 € / 100µg
Estimated Shipment: 12-09-2024
to see stock.

Description

Description

Specifications

Specifications

Galectin-13
Cytoplasmic and Cell Surface
PP13/1161
Unconjugated
XP_016882204
Mouse
16kDa (monomer) ;32kDa (homodimer)
Protein A/G Purified
RUO
29124
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
LGALS13
Recombinant human Galectin-13 protein fragment (aa23-134)
K-562 cells. Placenta or Spleen.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers