missing translation for 'onlineSavingsMsg'
Learn More

Galena, naturally occurring mineral, approximately 1-2in

Product Code. 15466485
Change view
Click to view available options
Quantity:
1 Ea.
5 Pc.
Unit Size:
Each
Pack of 5
Product Code. Quantity unitSize
15466485 1 Ea. Each
15476485 5 Pc. Pack of 5
2 options
This item is not returnable. View return policy

Product Code. 15466485

Brand: Thermo Scientific Alfa Aesar 042546.KF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-87-0 | PbS | 239.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name sulfanylidenelead
SMILES [S--].[Pb++]

Specifications

Physical Form Crystal Clusters
Quantity 1 Ea.
UN Number UN3077
Solubility Information Soluble in strong HNO3,in excess of hot HCl
Formula Weight 239.25
Packaging Bag
Chemical Name or Material Galena

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.