missing translation for 'onlineSavingsMsg'
Learn More

Galena, naturally occurring mineral, grains, approximately 0.06-0.19in

Product Code. 11385347
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385347 5 g 5g
11365347 1 g 1g
11375347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385347 Supplier Thermo Scientific Alfa Aesar Supplier No. 042640.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1314-87-0 | PbS | 239.30 g/mol

Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-87-0
Molecular Formula PbS
Molecular Weight (g/mol) 239.30
MDL Number MFCD00016280
InChI Key FILXVLQXZYXBSJ-UHFFFAOYSA-N
Synonym lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide
PubChem CID 14819
IUPAC Name sulfanylidenelead
SMILES [S--].[Pb++]

Specifications

Color Black
Physical Form Powder, Flake, Crystalline, Beads, Lump, Etc.
Quantity 5 g
Solubility Information Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide.
Formula Weight 239.25
Odor Odorless
Chemical Name or Material Galena, naturally occurring mineral, grains, approximately 0.06-0.19 in.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.