missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallamine Triethiodide, TRC
Specifications
Specifications
| Chemical Name or Material | Gallamine triethiodide |
| CAS | 65-29-2 |
| Molecular Formula | C30 H60 N3 O3 . 3 I |
| Quantity | 2.5 g |
| Synonym | Ammonium, [v-phenenyltris(oxyethylene)]tris[triethyl-, triiodide (8CI), Ethanaminium, 2,2',2''-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, triiodide (9CI), [v-Phenenyltris(oxyethylene)]tris[triethylammonium iodide] (6CI,7CI), 1,2,3-Tri(β-diethylaminoethoxy)benzene triethiodide, 1,2,3-Tris(2-diethylaminoethoxy)benzene triethiodide, 1,2,3-Tris(2-diethylaminoethoxy)benzene tris(ethyliodide), 1,2,3-Tris(2-triethylammonium ethoxy)benzene triiodide, 1,2,3-Tris(diethylaminoethoxy)benzene triethiodide, 3697 RP, Benzcurine iodide, F 2559, Flacedil, Flaxedil, Flaxedil iodide, Fourneau 2559, Gallaflex, Gallamin triethiodide, Gallamine iodide, Gallamine triethiodide, Gallamine triiodoethylate, HL 8583, Miowas G, Parexyl, Pyrogallol 1,2,3-(diethylaminoethyl ether) tris(ethyliodide), Pyrolaxon, RP 3697, Relaxan, Remyolan, Retensin, Sincurarine, Syncurarine, Tri(diethylaminoethoxy)-1,2,3-benzene triiodoethylate, Tri(β-diethylaminoethoxy)-1,2,3-benzene tri-iodoethylate, Ethanaminium, 2,2',2''-[1,2,3-benzenetriyltris(oxy)]tris[N,N,N-triethyl-, iodide (1:3) |
| InChI Formula | InChI=1S/C30H60N3O3.3HI/c1-10-31(11-2,12-3)22-25-34-28-20-19-21-29(35-26-23-32(13-4,14-5)15-6)30(28)36-27-24-33(16-7,17-8)18-9;;;/h19-21H,10-18,22-27H2,1-9H3;3*1H/q+3;;;/p-3 |
| SMILES | [I-].[I-].[I-].CC[N+](CC)(CC)CCOc1cccc(OCC[N+](CC)(CC)CC)c1OCC[N+](CC)(CC)CC |
| IUPAC Name | 2-[2,3-bis[2-(triethylazaniumyl)ethoxy]phenoxy]ethyl-triethylazanium;triiodide |
| Molecular Weight (g/mol) | 891.53 |
| Formula Weight | 891.1769 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?