missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid monohydrate, ACS reagent

Product Code. 13498288 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13498288 500 g 500g
13458618 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13498288 Supplier Thermo Scientific Acros Supplier No. 447585000
  • ONLINE EXCLUSIVE PRICE
    224.00 €
    207.00 € / 500g
    Save 17.00 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 251.0°C
Quantity 500 g
Packaging Plastic bottle
Linear Formula (HO)3C6H2CO2H·H2O
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, glycerol and acetone, practically insoluble in benzene, chloroform and, petroleum ether
IUPAC Name 3,4,5-trihydroxybenzoate
Formula Weight 188.12
Grade ACS Reagent
Chemical Name or Material Gallic acid monohydrate
compliance-icons
Identifiant produit
  • Gallic acid monohydrate
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contain: Gallic acid monohydrate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.