missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid monohydrate

Product Code. 10286270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10286270 100 g Plastic bottle 100g
10677362 500 g Plastic bottle 500g
10259493 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286270 Supplier Thermo Scientific Acros Supplier No. 220761000
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5995-86-8 | C7H5O5 | 169.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
IUPAC Name 3,4,5-trihydroxybenzoic acid;hydrate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 251.0°C
Color White to Beige
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 10% max. (120°C)
Packaging Plastic bottle
Physical Form Powder
Linear Formula (HO)3C6H2CO2H·H2O
Beilstein 10, 470
Merck Index 15, 4375
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: soluble in alcohol, glycerol and acetone, practically insoluble in benzene, chloroform and, petroleum ether
Formula Weight 188.12
Percent Purity ≥99% (on dry substance)
Chemical Name or Material Gallic acid monohydrate, c.p.
Show More Show Less
compliance-icons
Product Identifier
  • Gallic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Gallic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.