missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) isopropoxide, mixture of oligomers, 99%

Product Code. 11378018
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378018 5 g 5g
11388018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378018 Supplier Thermo Scientific Alfa Aesar Supplier No. 036583.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4452-61-3 | C3H8GaO | 129.819 g/mol

Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4452-61-3
Molecular Formula C3H8GaO
Molecular Weight (g/mol) 129.819
MDL Number MFCD00144389
InChI Key JTPVILBGRKEEOP-UHFFFAOYSA-N
Synonym 2-propanol, gallium salt
PubChem CID 122197675
IUPAC Name gallium;propan-2-ol
SMILES CC(C)O.[Ga]

Specifications

Melting Point 72°C
Color Yellow
Density 1.35 g/mL
Boiling Point 120°C (1 mm)
Flash Point 68°C (154°F)
Linear Formula Ga(OC3H7)3
Quantity 5 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 246.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Gallium(III) isopropoxide, Mixture of oligomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.