missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis)
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Tamaño de la unidad:
100g
1g
25g
5g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
Especificaciones
Especificaciones
| Melting Point | 110.0°C |
| Color | White |
| Physical Form | Crystalline Powder, Crystals and/or Chunks |
| Quantity | 1 g |
| Assay Percent Range | 99.9998% |
| Linear Formula | Ga(NO3)3·xH2O |
| Merck Index | 14, 4350 |
| Solubility Information | Solubility in water: soluble |
| IUPAC Name | gallium(3+) trinitrate |
| Formula Weight | 255.74 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Gallium(III) nitrate hydrate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido