Learn More
Gallium Indium Tin eutectic, 99.99% (metals basis)
Description
Gallium-Indium eutectic (EGAIn) is an electrically conductive fluid metal. It useful as a contact electrode for the electrical characterization of thin film organic and semiconductor devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Gallium Indium Tin eutectic |
| Melting Point | 10.7°C |
| Color | White |
| Physical Form | Liquid |
| Quantity | 1 g |
| Assay Percent Range | 99.99% (Metals basis) |
| Molecular Formula | Ga:In:Sn; 62:22:16 wt.% |
| MDL Number | MFCD00144388 |
| Solubility Information | Insoluble in water or organic solvents. |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.