missing translation for 'onlineSavingsMsg'
Learn More

Gallium Tin alloy, 99.99% (metals basis)

Product Code. 11393417
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393417 50 g 50g
11303427 1 g 1g
11383417 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393417 Supplier Thermo Scientific Alfa Aesar Supplier No. 018161.18
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5 , 7440-55-3
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver
Melting Point ≥25°C
Appearance Silver-colored liquid
Quantity 50 g
Assay Percent Range 99.99% (metals basis)
Form Melting Solid
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Tin: 8.0%; Gallium: 92.0%
Chemical Name or Material Gallium Tin alloy

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.