missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Galloflavin

Product Code. 30001522
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001522 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001522 Supplier Tocris Bioscience™ Supplier No. 4795/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lactate dehydrogenase inhibitor; impairs aerobic glycolysis

Inhibitor of human lactate dehydrogenase (LDH) (Ki values are 5.46 and 15.1 μM for LDH-A and LDH-B, respectively, in competition with pyruvate). Inhibits lactate production and decreases ATP synthesis in PLD/PRF/5 cells; thought to decrease cell viability via impairment of aerobic glycolysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 568-80-9
Quantity 10 mg
Molecular Formula C12H6O8
Formula Weight 316.26
Purity >95%
Molecular Weight (g/mol) 278.17
Chemical Name or Material Galloflavin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.