missing translation for 'onlineSavingsMsg'
Learn More

Manutan Galvanised wire rope, medium duty, 3000kg WLL capacity, 50m long coil

Product Code. 12828718
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12828718 Supplier Manutan Supplier No. A341479
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Galvanised wire rope manufactured to current British Standards. Medium duty rope ideal for lifting heavy loads. WLL based on 5:1 safety factor.

  • Diameter: 16mm
  • Roll length: 50m
  • WLL: 3000kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.