missing translation for 'onlineSavingsMsg'
Learn More

Gamma-Aminobutyric Acid, TRC

Product Code. 30009001
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009001 10 g 10g
30012342 1 g 1g
30004202 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009001 Supplier Toronto Research Chemicals Supplier No. TRCA60292010G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4 H9 N O2
Molecular Weight (g/mol) 103.12
Synonym Vigabatrin impurity D (EP), Gamma Aminobutyric Acid, 4-Aminobutanoic Acid, γ-Aminobutyric Acid, Butanoic acid, 4-amino-, Butyric acid, 4-amino- (7CI,8CI), 3-Carboxypropylamine, 4-Aminobutyric acid, Aminalon, GABA, Gaballon, Gamarex, Gammalon, Gammalone, Gammar, Gammasol, Mielogen, Mielomade, NSC 27418, NSC 32044, NSC 45460, NSC 51295, Oryza GABA Extract HC 90, Pharma-GABA 20S, Pharmagaba, Pharmagaba 20, Pharmagaba 20D, Piperidic acid, Piperidinic acid, γ-Aminobutanoic acid, ω-Aminobutyric acid
IUPAC Name 4-aminobutanoic acid
SMILES NCCCC(=O)O

Specifications

InChI Formula InChI=1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)
Quantity 10 g
Formula Weight 103.0633
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 4-Aminobutanoic acid (GABA)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.