missing translation for 'onlineSavingsMsg'
Learn More

gamma-Bisabolene (Mixture of Isomers) (Technical Grade), TRC

Product Code. 30138232
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138232 500 mg 500mg
30135864 1 g 1g
30011335 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138232 Supplier Toronto Research Chemicals Supplier No. TRCB399795500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-62-5
Molecular Formula C15H24
Molecular Weight (g/mol) 204.35
Synonym 2-Methyl-6-(4-methyl-3-cyclohexen-1-ylidene)-2-heptene, 4-(1,5-Dimethyl-4-hexen-1-ylidene)-1-methylcyclohexene
IUPAC Name 1-methyl-4-(6-methylhept-5-en-2-ylidene)cyclohexene
SMILES CC1=CC/C(CC1)=C(C)/CC/C=C(C)\C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H24/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8H,5,7,9-11H2,1-4H3
Formula Weight 204.1878
Chemical Name or Material gamma-Bisabolene (Mixture of Isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.