missing translation for 'onlineSavingsMsg'
Learn More

gamma-CEHC, TRC

Product Code. 30136818
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136818 25 mg 25mg
30136224 100 mg 100mg
30009521 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136818 Supplier Toronto Research Chemicals Supplier No. TRCC24920025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178167-75-4
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.32
Synonym 3,4-Dihydro-6-hydroxy-2,7,8-trimethyl-2H-1-benzopyran-2-propanoic Acid, (±)-LLU-α, 2,7,8-Trimethyl-2-(β-carboxyethyl)-6-hydroxychroman, Racemic LLU-α
IUPAC Name 3-(6-hydroxy-2,7,8-trimethyl-3,4-dihydrochromen-2-yl)propanoic acid
SMILES OC(=O)CCC1(C)CCc2cc(O)c(C)c(C)c2O1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H20O4/c1-9-10(2)14-11(8-12(9)16)4-6-15(3,19-14)7-5-13(17)18/h8,16H,4-7H2,1-3H3,(H,17,18)
Formula Weight 264.1362
Percent Purity >95%
Chemical Name or Material gamma-CEHC
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.