missing translation for 'onlineSavingsMsg'
Learn More

gamma-Decanolactone, 98%

Product Code. 11448617
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448617 25 g 25g
11468900 5 g 5g
11458617 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448617 Supplier Thermo Scientific Alfa Aesar Supplier No. A19732.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 706-14-9 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706-14-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00005404
InChI Key IFYYFLINQYPWGJ-UHFFFAOYNA-N
Synonym gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
PubChem CID 12813
IUPAC Name 5-hexyloxolan-2-one
SMILES CCCCCCC1CCC(=O)O1

Specifications

Density 0.948
Boiling Point 150°C to 153°C (15 mmHg)
Flash Point 146°C (294°F)
Refractive Index 1.5
Quantity 25 g
Beilstein 117547
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material gamma-Decanolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.