missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Gamma Irradiated Absorbent Towel, 4 Ply

Product Code. 18899816 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Quantity:
13720 Pk.
490 Pk.
Unit Size:
Pack of 13720
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18899816 490 Pk. -
18833617 13720 Pk. Pack of 13720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18899816 Supplier Keystone Cleanroom Products Supplier No. 770ST14_490PCS
Estimated Shipment: 15-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Higly Absorbent, Scrim Reinforced Sterile Towels.

  • Gamma Irradiated, nonwoven towels have a very high absorbency rate which allows for wiping jobs to be completed quickly
  • The product is made of multiple layers of high grade cellulose tissue with scrim reinforcing between the plies for added durability and good scrubbing action
  • Noted for being low linting and having good wipe dry properties

Specifications

Product Type Scrubber
Color-coded No
Handle Type None
Sterility Sterile
Autoclavable Autoclavable
Certifications/Compliance Certificate of Sterility
Color White
Disposable Single-use
For Use With (Application) Wiping
Length (English) 18 in.
Quantity 490 Pk.
Width (English) 18 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.