Learn More
Keystone Cleanroom Products Gamma-Irradiated Bic Pen
Product Code. 17776899 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17776899 Black Pack of 500
17786899 Blue Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17776899 Supplier Keystone Cleanroom Products Supplier No. S01PENBLK
  • ONLINE EXCLUSIVE PRICE
    1360.00 €
    1106.94 € / Pack of 500
    Save 253.06 €
    18% Off
Estimated Shipment: 18-06-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Product Type Cleanroom Pen
Color Black
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation
No. per Case 500/Cs.
Packaging Type Peel Pouch
Length (Metric) 14 cm