Learn More
Keystone Cleanroom Products Gamma-Irradiated Ultra Fine Point Sharpie
Product Code. 17716909 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17716909 Black Pack of 500
17726909 Blue Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17716909 Supplier Keystone Cleanroom Products Supplier No. S01UFMARKBLK
Only null left
Add to basket
Add to basket
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Tip Style Ultrafine
Product Type Cleanroom Marker
Color Black
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation