missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Gamma Maxi Radiation Bins

Product Code. 12357127 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
164mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12357127 164mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12357127
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This gamma radiation bin, designed to hold up to three midi-box inserts, is ideal for holding microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.

  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased

Specifications

For Use With (Application) Designed to hold up to three midi-box inserts
Height (Metric) 164mm
Product Type Radiation Bin
Width (Metric) 304mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.