missing translation for 'onlineSavingsMsg'
Learn More

gamma-Octanolactone, 98%

Product Code. 11423657
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423657 25 g 25g
11433657 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423657 Supplier Thermo Scientific Alfa Aesar Supplier No. A18142.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-50-7 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-50-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00005402
InChI Key IPBFYZQJXZJBFQ-UHFFFAOYSA-N
Synonym gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
PubChem CID 7704
IUPAC Name 5-butyloxolan-2-one
SMILES CCCCC1CCC(=O)O1

Specifications

Density 0.981
Boiling Point 233°C to 235°C
Flash Point 126°C (259°F)
Refractive Index 1.444
Quantity 25 g
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material gamma-Octanolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.