missing translation for 'onlineSavingsMsg'
Learn More

gamma-Phenyl-gamma-butyrolactone, 97%

Product Code. 11485797
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485797 25 g 25g
11475797 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485797 Supplier Thermo Scientific Alfa Aesar Supplier No. B24364.14
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1008-76-0 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-76-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00005398
InChI Key AEUULUMEYIPECD-UHFFFAOYNA-N
Synonym gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
PubChem CID 13884
IUPAC Name 5-phenyloxolan-2-one
SMILES O=C1CCC(O1)C1=CC=CC=C1

Specifications

Melting Point 35°C to 38°C
Density 1.155
Boiling Point 305°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 125169
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material gamma-Phenyl-gamma-butyrolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.