missing translation for 'onlineSavingsMsg'
Learn More

gamma-Valerolactone, 98%

Product Code. 11423128
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423128 500 g 500g
11458900 25 g 25g
11413128 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423128 Supplier Thermo Scientific Alfa Aesar Supplier No. A18872.36
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-29-2 | C5H8O2 | 100.117 g/mol

It finds it application as a food additive that is used to improve the taste or odor of a food. It is also used as toiletry fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a food additive that is used to improve the taste or odor of a food. It is also used as toiletry fragrances.

Solubility
Fully miscible in water (9.381e+004 mg/L @ 25°C (est.)). Soluble in alcohol, fixed oils.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-29-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00005400
InChI Key GAEKPEKOJKCEMS-UHFFFAOYSA-N
Synonym gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
PubChem CID 7921
ChEBI CHEBI:48569
IUPAC Name 5-methyloxolan-2-one
SMILES CC1CCC(=O)O1

Specifications

Melting Point -31°C
Density 1.052
Boiling Point 207°C to 208°C
Flash Point 81°C (178°F)
Odor Pleasant
Refractive Index 1.433
Quantity 500 g
Beilstein 80420
Solubility Information Fully miscible in water (9.381e+004mg/L @ 25°C (est.)). Soluble in alcohol,fixed oils.
Formula Weight 100.12
Percent Purity ≥98%
Chemical Name or Material gamma-Valerolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.