missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GammaBind™ G Sepharose™ Medium

Product Code. 11574945 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
5 mL
25 mL
250 mL
Unit Size:
250mL
25mL
5mL
Product Code. Volume (Metric) unitSize
11574945 250 mL 250mL
11554945 5 mL 5mL
11564945 25 mL 25mL
3 options
This item is not returnable. View return policy

Product Code. 11574945

Brand: Cytiva 17088504

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use recombinant GammaBind™ for laboratory scale antibody purification. Cytiva Life Sciences™ GammaBind™ G Sepharose™ Medium binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

Binds to all IgG subclasses from human, mouse, and rat and total IgG from guinea pig, rabbit, goat, cow, sheep, and horse. Engineered for antibody purification at laboratory scale.

  • Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin
  • Ligand: GammaBind G type 2 lacking albumin- binding region
  • Ligand coupling method: cyanogen bromide activation
  • Ligand density: ≈ 3mg GammaBind G, type 2/mL medium
  • Number of IgG binding sites per ligand: 2
  • Matrix: Agarose, 4%
  • Average particle size: 90μm
  • Binding capacity: >18 mg human IgG/ml, 6 mg mouse IgG/ml
  • pH stability: 3-9 (working), 2-9 (short term)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sepharose Medium
Binding Capacity >18 mg/mL Human IgG, 6 mg/mL Mouse IgG
Ligand Concentration ≈3 mg GammaBind G, Type 2/mL drained gel
Particle Size 45 to 165 μm
pH 3 to 9
Volume (Metric) 250 mL
For Use With (Application) For protein purification by affinity coupling
Storage Buffer PBS pH 7, 20% Ethanol
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.