missing translation for 'onlineSavingsMsg'
Learn More

Ganciclovir, 98%, Thermo Scientific Chemicals

Product Code. 15701779 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15701779 1 g 1g
15711779 5 g 5g
15721779 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15701779 Supplier Thermo Scientific Acros Supplier No. 461710010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82410-32-0
Molecular Formula C9H13N5O4
Molecular Weight (g/mol) 255.23
InChI Key IRSCQMHQWWYFCW-UHFFFAOYSA-N
Synonym ganciclovir, gancyclovir, cytovene, vitrasert, hydroxyacyclovir, ganciclovirum, zirgan, ganciclovir sodium, cymevene, hhemg
PubChem CID 3454
ChEBI CHEBI:465284
IUPAC Name 2-amino-9-(1,3-dihydroxypropan-2-yloxymethyl)-3H-purin-6-one
SMILES C1=NC2=C(N1COC(CO)CO)NC(=NC2=O)N

Specifications

Melting Point 251.0°C to 252.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 3,14g/L (25°C).
Formula Weight 255.23
Percent Purity 98%
Chemical Name or Material Ganciclovir
compliance-icons
Product Identifier
  • Ganciclovir
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity on or via lactation
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H351-Suspected of causing cancer.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ganciclovir sodium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.