missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Ganciclovir ReadyMade™ Solution

Product Code. 30044031
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044031 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044031 Supplier TOKU-E™ Supplier No. G09310x1ML
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ganciclovir ReadyMade™ Solution is provided as a sterile-filtered solution in DMSO.

  • CAS Number: 82410-32-0
  • Molecular Formula: C9H13N5O4
  • Molecular Weight: 255.23 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Synthetic
  • Spectrum: Ganciclovir is an antiviral compound specifically used to treat cytomegalovirus (CMV) in immunocompromised individuals
  • Mechanism of Action: Ganciclovir is first phosphorylated to Ganciclovir monophosphate by a viral kinase encoded by the cytomegalovirus (CMV) gene UL97. After that, cellular kinases convert this molecule to ganciclovir-5'-triphosphate which specifically targets cytomegalovirus (CMV) DNA polymerase since it is a competitive inhibitor of dGTP thus it gets incorporated into DNA and inhibits viral DNA polymerases more than the cellular DNA polymerases, resulting in termination of viral DNA elongation

Specifications

Chemical Name or Material Ganciclovir
CAS 82410-32-0
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Product Line ReadyMade™ Solutions
Target CMV, viral DNA elongation
Molecular Formula C9H13N5O4
Formula Weight 255.23 amu
Physical Form Liquid
Color Colorless
Product Type Antibiotic
Purity >99%
Molecular Weight (g/mol) 255.23 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.