missing translation for 'onlineSavingsMsg'
Learn More

Ganciclovir, TRC

Product Code. 18083130
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083130 1 g 1g
18050220 50 mg 50mg
17949739 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083130 Supplier Toronto Research Chemicals Supplier No. TRCG2350001G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82410-32-0
Molecular Formula C9 H13 N5 O4
Molecular Weight (g/mol) 255.23
Synonym Ganciclovir, 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-, 2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 2'-NDG, 2'-Nor-2'-deoxyguanosine, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, BW 759, BW 759U, BW-B 759U, Biolf 62, DHPG, Denosine, Gancyclovir, Ganguard, Ganquard, Gengxiluowei, HHEMG, Hydroxyacyclovir, Nataclovir, Natclovir, RS 21592, Vitrasert, Valganciclovir Hydrochloride Imp. A (EP)
IUPAC Name 2-amino-9-(1,3-dihydroxypropan-2-yloxymethyl)-1H-purin-6-one
SMILES NC1=Nc2c(ncn2COC(CO)CO)C(=O)N1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)
Formula Weight 255.0968
Percent Purity >95%
Chemical Name or Material Ganciclovir
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.