Learn More
Paragon Scientific™ Gardner (ASTM D1544, D6166) Colour Reference Standard
Gardner Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
Brand:  Paragon Scientific™ 134200
Product Code. 13585330
  • 240.00 € / 500mL
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to International Standards
  • 500mL Pack Size supplied with MSDS
  • 18 months shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom