Learn More
Thermo Scientific™ Gas Chromatography SMART Syringe for TriPlus™ RSH SMART Autosampler
Shop All Thermo Scientific ProductsDescription
Use the unique technology of Thermo Scientific™ gas chromatography SMART Syringes as a traceable usage-based approach to gas chromatography (gas chromatography) syringe management, resulting in increased reliability, instrument up-time, confidence in the results, and full traceability.
SMART Technology Benefits:
- Gain higher confidence in analytical results
- Ensure error-free usage of syringes through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring usage conditions and lifetime
- Facilitate syringe management with a color-coded health notification dashboard integrated in Chromeleon™ CDS
Specifications
Specifications
| Material | Borosilicate Glass (Barrel) |
| Product Type | Syringe |
| Volume (Metric) | 10 μL |
| Graduated | Yes |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.