Learn More
Macherey-Nagel™ Replacement Gaskets for NucleoVac 96
Product Code. 15829067
Change view
Click to view available options
Quantity:
2 Each
Unit Size:
1 set
Product Code. Quantity unitSize
15829067 2 Each 1 set
1 options
Product Code. 15829067 Supplier Macherey-Nagel™ Supplier No. 740248
In Stock
Add to basket
Add to basket
Set of two gaskets for the NucleoVac 96 Vacuum Manifold
The NucleoVac 96 Vacuum Manifold is designed for the rapid manual parallel purification of nucleic acids using the 8-well and 96-well NucleoSpin and NucleoFast kits. Time consuming centrifugation and decanting steps are eliminated by use of vacuum.
  • NucleoVac 96 Vacuum Manifold consists of the manifold base and lid, a spacer set, and a waste container set.
  • Spacer set includes:
    • 2 spacers MTP/MULTI-96 PLATES
    • 2 spacers ROUND-WELL BLOCKS
    • 2 spacers SQUARE-WELL BLOCKS
    • 2 spacers MICROTUBE RACKS
  • For use of NucleoSpin™ 8‐well strips, Starter Set A (column holder and dummy strips) is required
Specifications
Product Type Gaskets
For Use With (Equipment) NucleoVac 96 Vacuum Manifold
Includes Set of replacement gaskets
Quantity 2 Each