missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Gasket Valve

Product Code. 15318467 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone™ freeze dryer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15318467 FreeZone™ freeze dryer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15318467 Supplier Labconco™ Supplier No. 7505800
  • ONLINE EXCLUSIVE PRICE
    2.81 €
    2.07 € / Pack of 1
    Save 0.74 €
    26% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Labconco™ Gasket Valve is a replacement rubber gasket for use with FreeZone freeze dryers.

FreeZone Freeze Dryers

  • Designed to handle the lyophilization needs of research and pilot plant laboratories
  • All sizes/models include a large touch screen display
  • Lyo-Works™ OS
    • Provides a real time display of temperature and vacuum level
    • Optional End-Zone™ end point detection system
  • Console models are offered in a range of sizes with optional stoppering tray
  • Collectors that reach -50°C are suitable for lyophilizing aqueous samples
  • Collectors that reach -84°C are ideal for freeze drying samples with low eutectic temperatures, including ones containing acetonitrile

Specifications

Type Gasket
Color Black
Model 7505800
Material Rubber
For Use With (Equipment) FreeZone™ freeze dryer
Weight (English) 1.0 lb.
Weight (Metric) 0.5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.